Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TATA binding protein TBP Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB318352 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318352 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318352 Supplier Novus Biologicals Supplier No. NBP298905JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TATA binding protein TBP Polyclonal antibody specifically detects TATA binding protein TBP in Human samples. It is validated for Western Blot

Specifications

Antigen TATA binding protein TBP
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human TATA binding protein TBP.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6908
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.