Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TATA binding protein TBP Antibody (SR1605), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32166550UL
Description
TATA binding protein TBP Monoclonal antibody specifically detects TATA binding protein TBP in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TATA binding protein TBP | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1605 | |
| Western Blot 1:1000-1:2000 | |
| GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit | |
| A synthesized peptide derived from human TATA binding protein TBP (Uniprot #: P20226) | |
| 50 μg | |
| Transcription Factors and Regulators | |
| 6908 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction