Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TATA binding protein TBP Antibody (SR1605), Novus Biologicals™
SDP

Catalog No. p-200006628 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259603 100 μg
NB259478 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259603 Supplier Novus Biologicals Supplier No. NBP321665100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TATA binding protein TBP Monoclonal antibody specifically detects TATA binding protein TBP in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen TATA binding protein TBP
Applications Western Blot
Classification Monoclonal
Clone SR1605
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit
Host Species Rabbit
Immunogen A synthesized peptide derived from human TATA binding protein TBP (Uniprot #: P20226)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6908
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.