Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Antibody (2E9), Novus Biologicals™
SDP

Catalog No. NBP225162SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225162SS 0.025 mL
NBP225162 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225162SS Supplier Novus Biologicals Supplier No. NBP225162SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tau Monoclonal specifically detects Tau in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Tau
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2000000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Accession No. P10636
Gene Alias DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau
Gene Symbols MAPT
Host Species Mouse
Immunogen Recombinant full length 441 amino acid human tau. Epitope located in the peptide KDRVQSKIGSLDNITHVPGG, amino acids 347-366.
Molecular Weight of Antigen 48-67 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cytokine Research, Cytoskeleton Markers, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4137
Test Specificity This antibody is expected to bind to all Tau isoforms
Target Species Human, Mouse, Rat, Pig, Bovine, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.