Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Antibody (Tau-5), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB015851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015851 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015851 Supplier Novus Biologicals Supplier No. NBP281091AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Tau Monoclonal specifically detects Tau in Human, Mouse, Rat, Sheep samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Tau
Applications Immunofluorescence
Classification Monoclonal
Clone Tau-5
Conjugate Alexa Fluor 647
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau
Gene Symbols MAPT
Host Species Rabbit
Immunogen Tau protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4137
Test Specificity This antibody binds to sequence 210-241 of tau protein.
Target Species Human, Mouse, Rat, Sheep
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.