Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Oligomeric Complex Antibody (TOC1), Alexa Fluor™ 594 - Chimeric, Novus Biologicals™
SDP

Catalog No. NB236020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB236020 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB236020 Supplier Novus Biologicals Supplier No. NBP320162AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Tau Oligomeric Complex Monoclonal antibody specifically detects Tau Oligomeric Complex in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Tau Oligomeric Complex
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone TOC1
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Tau Oligomeric Complex 1, TOC1, TOC-1
Host Species Rabbit
Immunogen The original antibody was generated by immunizing a mouse with electro eluted cross-linked Tau dimers.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.