Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau, p Ser199 Antibody (23E8), Novus Biologicals™
SDP

Catalog No. NB298154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298154 50 μL
NB298155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298154 Supplier Novus Biologicals Supplier No. NBP32641150UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Tau Monoclonal antibody specifically detects Tau in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry

Specifications

Antigen Tau
Applications Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 23E8
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Tau [p Ser199] [UniProt P10636]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4137
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.