Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tau, p Ser396 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$246.40 - $418.55
Specifications
| Antigen | Tau (p Ser396) |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017886
![]() |
Novus Biologicals
NB100822430.025ML |
0.025 mL |
Each for $246.40
|
|
|||||
NB10082243
![]() |
Novus Biologicals
NB10082243 |
0.05 mL |
Each for $418.55
|
|
|||||
Description
Tau Polyclonal specifically detects Tau in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Tau (p Ser396) | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Rat, Human, Mouse | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| MAPT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cytokine Research, Cytoskeleton Markers, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers | |
| P10636 | |
| 4137 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title