Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau, p Ser396 Antibody, Novus Biologicals™
SDP

Catalog No. NB10082243 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10082243 0.05 mL
NB017886 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10082243 Supplier Novus Biologicals Supplier No. NB10082243
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tau Polyclonal specifically detects Tau in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Tau (p Ser396)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:100
Formulation PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide
Gene Accession No. P10636
Gene Alias DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau
Gene Symbols MAPT
Host Species Rabbit
Immunogen This Tau [p Ser396] Antibody was produced against synthesized phosphopeptide derived from human Tau around the phosphorylation site of serine 396 (Y-K-SP-P-V).
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cytokine Research, Cytoskeleton Markers, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4137
Test Specificity Detects endogenous levels of Tau only when phosphorylated at serine396.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.