Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau, p Thr205, p Ser202 Antibody (AH36), Novus Biologicals™
SDP

Catalog No. NB171889 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171889 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171889 Supplier Novus Biologicals Supplier No. NBP318265
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Tau Monoclonal antibody specifically detects Tau in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Dot Blot

Specifications

Antigen Tau
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Dot Blot
Classification Monoclonal
Clone AH36
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500, Dot Blot
Formulation PBS pH 7.4, 50% glycerol
Gene Alias DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau
Host Species Rabbit
Immunogen Synthetic peptide of Human Phospho Tau (Ser202/Thr205)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4137
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.