Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurine, 99%

Catalog No. p-3346554
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 107-35-7 | C2H7NO3S | 125.14 g/mol

Taurine plays an important role in cardiovascular function, skeletal muscle function, the retina and the central nervous system. It acts as an antioxidant as well as protecting against the toxicity of various substances such as lead and cadmium. It is also used in contact lens solution and an ingredient in numerous energy drinks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Taurine plays an important role in cardiovascular function, skeletal muscle function, the retina and the central nervous system. It acts as an antioxidant as well as protecting against the toxicity of various substances such as lead and cadmium. It is also used in contact lens solution and an ingredient in numerous energy drinks.

Solubility
Soluble in water and dimethyl sulfoxide. Insoluble in absolute ethanol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-35-7
Molecular Formula C2H7NO3S
Molecular Weight (g/mol) 125.14
MDL Number MFCD00008197
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
Synonym taurine, tauphon, ethanesulfonic acid, 2-amino, l-taurine, 2-aminoethylsulfonic acid, o-due, 2-sulfoethylamine, taufon, aminoethanesulfonic acid, aminoethylsulfonic acid
PubChem CID 1123
ChEBI CHEBI:15891
IUPAC Name 2-aminoethanesulfonic acid
SMILES NCCS(O)(=O)=O

Specifications

Melting Point >300°C
Assay Percent Range 99%
Quantity 100 g
Beilstein 1751215
Solubility Information Soluble in water and dimethyl sulfoxide. Insoluble in absolute ethanol and diethyl ether.
Formula Weight 125.15
Chemical Name or Material Taurine
EINECSNumber 203-483-8
RTECSNumber WX0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.