Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAX1BP1 Rabbit anti-Human, Mouse, Rat, Clone: 7Y9W3, Novus Biologicals™
SDP

Catalog No. NB166684 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166684 20 μg
NB166683 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166684 Supplier Novus Biologicals Supplier No. NBP31579420UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TAX1BP1 Monoclonal antibody specifically detects TAX1BP1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen TAX1BP1
Applications Western Blot
Classification Monoclonal
Clone 7Y9W3
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias T6BP, Tax1 (human T-cell leukemia virus type I) binding protein 1, tax1-binding protein 1, TRAF6-binding protein, TXBP151CALCOCO3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 690-789 of human TAX1BP1 (Q86VP1). DSEDSKEDENVPTAPDPPSQHLRGHGTGFCFDSSFDVHKKCPLCELMFPPNYDQSKFEEHVESHWKVCPMCSEQFPPDYDQQVFERHVQTHFDQNVLNFD
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8887
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.