Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taxol Resistant Associated Protein 3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP266585 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266585 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266585 Supplier Novus Biologicals Supplier No. NBP266585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Taxol Resistant Associated Protein 3 Polyclonal specifically detects Taxol Resistant Associated Protein 3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen TaxolResistantAssociatedProtein3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, ELISA 1:100-1:2000
Formulation PBS with 0.08% Sodium Azide
Gene Accession No. Q9Y5P2
Gene Alias TRAG3
Host Species Rabbit
Immunogen Synthetic peptide derived from the human Trag-3 protein.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.