Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Taxol
GREENER_CHOICE

Catalog No. p-4785656
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Promotes assembly and inhibits disassembly of microtubules

Taxol, also known as paclitaxel, promotes and stabilizes tubulin polymerization, causing cell cycle arrest. In CCRF-HSB-2 cells, Taxol induces autocatalytic activation of caspase-10, triggering apoptosis. Taxol is an antitumor agent. In a mouse model of bladder cancer Taxol decreases tumor growth. Taxol also prolongs survival of mice transplanted with human ovarian carcinoma xenografts. Taxol improves neurological outcome and magnetic resonance imaging biomarkers after traumatic brain injury in mice.

Chemical Identifiers

CAS 33069-62-4
Molecular Formula C47H51NO14

Specifications

Quantity 10 mg
Formula Weight 853.92
Purity >99%
Molecular Weight (g/mol) 853.92
Chemical Name or Material Taxol
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.