Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAZ/WWTR1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications
Supplier: Novus Biologicals NB600220SS
Description
TAZ/WWTR1 Polyclonal specifically detects TAZ/WWTR1 in Human, Mouse samples. It is validated for Western Blot.Specifications
TAZ/WWTR1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1 | |
Rabbit | |
44.1 kDa | |
0.025 mg | |
Signal Transduction, Transcription Factors and Regulators | |
25937 | |
Mouse, Human | |
IgG |
Western Blot | |
1 mg/mL | |
Western Blot 1:500 -1:1000 | |
Q9GZV5 | |
WWTR1 | |
Synthetic peptide representing residues 386-400 of the human TAZ protein. | |
Affinity Purified | |
RUO | |
Primary | |
Reacts with residues 386-400, which is the PDZ-binding domain of the 49kDa human TAZ protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction