Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAZ/WWTR1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications
$206.50 - $499.50
Specifications
Antigen | TAZ/WWTR1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 -1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
TAZ/WWTR1 Polyclonal specifically detects TAZ/WWTR1 in Human, Mouse samples. It is validated for Western Blot.Specifications
TAZ/WWTR1 | |
Western Blot 1:500 -1:1000 | |
Polyclonal | |
Rabbit | |
Signal Transduction, Transcription Factors and Regulators | |
PBS with 0.02% Sodium Azide | |
FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1 | |
WWTR1 | |
IgG | |
Affinity Purified | |
Reacts with residues 386-400, which is the PDZ-binding domain of the 49kDa human TAZ protein. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse, Human | |
Q9GZV5 | |
25937 | |
Synthetic peptide representing residues 386-400 of the human TAZ protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
44.1 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title