Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAZ/WWTR1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11058359T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11058359T 0.025 mL
NB11058359 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11058359T Supplier Novus Biologicals Supplier No. NB11058359SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 11 publications

TAZ/WWTR1 Polyclonal specifically detects TAZ/WWTR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay.

Specifications

Antigen TAZ/WWTR1
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In situ PLA
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Simple Western 1:100, Chromatin Immunoprecipitation reported in scientific literature (PMID 25587023), Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500. Use reported in scientific literature (PMID 25587023), Proximity Ligation Assay reported in scientific literature (PMID 25587023), Chromatin Immunoprecipitation (ChIP)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9GZV5
Gene Alias FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1
Gene Symbols WWTR1
Host Species Rabbit
Immunogen Synthetic peptides made to the human TAZ protein. These peptides were selected for the lack of homology to YAP protein. [UniProt# Q9GZV5]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 25937
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.