Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAZ/WWTR1 Antibody (OTI1H9), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP274449G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274449G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274449G Supplier Novus Biologicals Supplier No. NBP274449G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAZ/WWTR1 Monoclonal specifically detects TAZ/WWTR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TAZ/WWTR1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1H9
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1
Gene Symbols WWTR1
Host Species Mouse
Immunogen Full length recombinant protein of human TAZ/WWTR1 (NP_057287) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Chromatin Research, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 25937.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.