Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TB 21007
GREENER_CHOICE

Catalog No. 290510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-051-0 10 mg
29-055-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-051-0 Supplier Tocris Bioscience™ Supplier No. 2905/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

α5-selective GABAA inverse agonist

GABAA receptor inverse agonist selective for the α5-subtype (Ki values are 1.6, 16, 20 and 20 nM for α5, α2, α1 and α3 subtypes respectively). Brain penetrant; enhances cognitive performance in rats in the delayed matching-to-place morris water maze test following i.p. administration.

Chemical Identifiers

CAS 207306-50-1
Molecular Formula C15H17NO2S3

Specifications

Quantity 10 mg
Formula Weight 339.5
Purity >99%
Molecular Weight (g/mol) 339.486
Chemical Name or Material TB 21007
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.