Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TBB
GREENER_CHOICE

Catalog No. 227510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-751-0 10 mg
22-755-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-751-0 Supplier Tocris Bioscience™ Supplier No. 2275/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective cell-permeable CK2 inhibitor

A cell-permeable, selective inhibitor of casein kinase-2 (CK2) (IC50 = 0.9 and 1.6 μM for rat liver and human recombinant CK2 respectively). Exhibits modest discrimination between CK2 subunits, with Ki values ranging from 80 nM to 210 nM. Acts in an ATP/GTP-competitive manner and displays one to two orders of magnitude selectivity over a panel of 33 protein kinases.

Chemical Identifiers

CAS 17374-26-4
Molecular Formula C6HBr4N3

Specifications

Quantity 10 mg
Formula Weight 434.71
Purity 98%
Molecular Weight (g/mol) 434.711
Chemical Name or Material TBB
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.