Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D10A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179843
Description
TBC1D10A Polyclonal specifically detects TBC1D10A in Human samples. It is validated for Western Blot.Specifications
TBC1D10A | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EBP50-PDX interactor of 64 kDa, EPI64, rab27A-GAP-alpha, TBC1 domain family member 10A, TBC1 domain family, member 10A | |
Rabbit | |
57 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%. | |
Human, Guinea Pig | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_114143 | |
TBC1D10A | |
Synthetic peptide directed towards the C terminal of human TBC1D10AThe immunogen for this antibody is TBC1D10A (NP_114143). Peptide sequence: KAILDAEPGPRPALQPSPSIRLPLDAPLPGSKAKPKPPKQAQKEQRKQMK | |
Affinity purified | |
RUO | |
83874 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction