Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D10A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TBC1D10A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TBC1D10A Polyclonal specifically detects TBC1D10A in Human samples. It is validated for Western Blot.Specifications
TBC1D10A | |
Polyclonal | |
Rabbit | |
NP_114143 | |
83874 | |
Synthetic peptide directed towards the C terminal of human TBC1D10AThe immunogen for this antibody is TBC1D10A (NP_114143). Peptide sequence: KAILDAEPGPRPALQPSPSIRLPLDAPLPGSKAKPKPPKQAQKEQRKQMK | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EBP50-PDX interactor of 64 kDa, EPI64, rab27A-GAP-alpha, TBC1 domain family member 10A, TBC1 domain family, member 10A | |
TBC1D10A | |
IgG | |
57 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title