Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP179030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP179030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP179030 Supplier Novus Biologicals Supplier No. NBP179030
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBC1D2 Polyclonal specifically detects TBC1D2 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen TBC1D2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-10ug/mg
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. EAW58884.1
Gene Alias Armus, FLJ10702, FLJ16244, FLJ42782, PARIS1member 2A, PP8997, TBC1 domain family, member 2
Gene Symbols TBC1D2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 878 and 928 of human TBC1 Domain Family, Member 2 using the numbering given in entry EAW58884.1 (GeneID 55357).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55357
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.