Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D21 Antibody (OTI2A6), PE, Novus Biologicals™
SDP

Catalog No. NBP274451PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274451PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274451PE Supplier Novus Biologicals Supplier No. NBP274451PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TBC1D21 Monoclonal specifically detects TBC1D21 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen TBC1D21
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2A6
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias MGC34741, TBC1 domain family member 21, TBC1 domain family, member 21
Gene Symbols TBC1D21
Host Species Mouse
Immunogen Full length recombinant protein of human TBC1D21 (NP_699187) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 161514.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.