Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D28 Antibody (OTI4A9), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274461B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274461B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274461B Supplier Novus Biologicals Supplier No. NBP274461B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TBC1D28 Monoclonal specifically detects TBC1D28 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TBC1D28
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4A9
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ40244, MGC138217, MGC138221, TBC1 domain family member 28, TBC1 domain family, member 28
Gene Symbols TBC1D28
Host Species Mouse
Immunogen Full length recombinant protein of human TBC1D28 (NP_001034486) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 254272.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.