Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017246 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017246 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017246 Supplier Novus Biologicals Supplier No. NBP281935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBC1D4 Polyclonal specifically detects TBC1D4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TBC1D4
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 10 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4
Gene Symbols TBC1D4
Host Species Rabbit
Immunogen TBC1D4 antibody was raised against an 19 amino acid peptide near the carboxy terminus of human TBC1D4 . The immunogen is located within amino acids 1190 - 1240 of TBC1D4. Amino Acid Squence: ETLEKLERANSQLKRQNMD
Molecular Weight of Antigen 140 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Diabetes Research, Lipid and Metabolism, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9882
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.