Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | TBC1D4 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TBC1D4 Polyclonal specifically detects TBC1D4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TBC1D4 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
O60343 | |
9882 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MEPPSCIQDEPFPHPLEPEPGVSAQPGPGKPSDKRFRLWYVGGSCLDHRTTLPMLPWLMAEIRRRSQKPE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Diabetes Research, Lipid and Metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4 | |
TBC1D4 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title