Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D4 Antibody (OTI5E6), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N274462PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274462PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274462PEC5 Supplier Novus Biologicals Supplier No. NBP274462PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TBC1D4 Monoclonal specifically detects TBC1D4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen TBC1D4
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5E6
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4
Gene Symbols TBC1D4
Host Species Mouse
Immunogen Full length recombinant protein of human TBC1D4 (NP_0055647) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Diabetes Research, Lipid and Metabolism, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 9882.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.