Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D9 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291866 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291866 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291866 Supplier Novus Biologicals Supplier No. NB10055321PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBC1D9 Polyclonal antibody specifically detects TBC1D9 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen TBC1D9
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias KIAA0882TBC1 domain family member 9, MDR1, TBC1 domain family member 9A, TBC1 domain family, member 9, TBC1 domain family, member 9 (with GRAM domain), TBC1D9A
Host Species Rabbit
Immunogen Maps to a region between residue 1118 and the C-terminus (residue 1168) of human TBC1 domain family, member 9 using the numbering given in entry NP_055945.1 (GeneID 23158).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23158
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.