Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBCC Antibody (OTI1B2), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP27446A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27446A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27446A94 Supplier Novus Biologicals Supplier No. NBP274465AF594

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

TBCC Monoclonal specifically detects TBCC in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot.

Specifications

Antigen TBCC
Applications Western Blot
Classification Monoclonal
Clone OTI1B2
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CFCtubulin-specific chaperone c, tubulin folding cofactor C, Tubulin-folding cofactor C
Gene Symbols TBCC
Host Species Mouse
Immunogen Full length recombinant protein of human TBCC (NP_003183) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6903.0
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.