Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBCC Antibody (OTI4C4), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N274467J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274467J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274467J646 Supplier Novus Biologicals Supplier No. NBP274467JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TBCC Monoclonal specifically detects TBCC in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TBCC
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4C4
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CFCtubulin-specific chaperone c, tubulin folding cofactor C, Tubulin-folding cofactor C
Gene Symbols TBCC
Host Species Mouse
Immunogen Full length recombinant protein of human TBCC (NP_003183) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6903.0
Target Species Human, Mouse, Rat, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.