Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBCCD1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15539420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15539420 20 μL
NBP155394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15539420 Supplier Novus Biologicals Supplier No. NBP15539420UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBCCD1 Polyclonal specifically detects TBCCD1 in Human samples. It is validated for Western Blot.

Specifications

Antigen TBCCD1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9NVR7
Gene Alias FLJ10560, TBCC domain containing 1, TBCC domain-containing protein 1
Gene Symbols TBCCD1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to TBCCD1(TBCC domain containing 1) The peptide sequence was selected from the N terminal of TBCCD1. Peptide sequence WPSPRNKSQSPDLTEKSNCHNKNWNDYSHQAFVYDHLSDLLELLLDPKQL.
Molecular Weight of Antigen 63 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55171
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.