Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBCD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | TBCD |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17991320
![]() |
Novus Biologicals
NBP17991320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179913
![]() |
Novus Biologicals
NBP179913 |
100 μL |
Each for $487.50
|
|
|||||
Description
TBCD Polyclonal specifically detects TBCD in Human samples. It is validated for Western Blot.Specifications
TBCD | |
Polyclonal | |
Rabbit | |
Human | |
Beta-tubulin cofactor D, KIAA0988, SSD1, SSD-1, TFCD, tubulin folding cofactor D, Tubulin-folding cofactor D, tubulin-specific chaperone d | |
TBCD | |
IgG | |
131 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_005984 | |
6904 | |
The immunogen for this antibody is TBCD. Peptide sequence KLVQRLGLTFLKPKVAAWRYQRGCRSLAANLQLLTQGQSEQKPLILTEDD. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title