Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBCK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TBCK |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156343
![]() |
Novus Biologicals
NBP156343 |
100 μL |
Each for $487.50
|
|
|||||
NBP15634320
![]() |
Novus Biologicals
NBP15634320UL |
20 μL | N/A | N/A | N/A | ||||
Description
TBCK Polyclonal specifically detects TBCK in Human samples. It is validated for Western Blot.Specifications
TBCK | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
HSPC302, MGC16169, TBC domain-containing protein kinase-like protein, TBC1 domain containing kinase, TBCKL | |
TBCK | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q8TEA7-3 | |
93627 | |
Synthetic peptides corresponding to MGC16169 The peptide sequence was selected from the middle region of MGC16169. Peptide sequence PPICTLPNFLFEDGESFGQGRDRSSLLDDTTVTLSLCQLRNRLKDVGGEA. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title