Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBK1 Antibody (108A429) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056705 0.1 mg
NB10056705T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056705 Supplier Novus Biologicals Supplier No. NB10056705
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 27 publications

TBK1 Monoclonal specifically detects TBK1 in Bovine, Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen TBK1
Applications Flow Cytometry, Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot
Classification Monoclonal
Clone 108A429
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Simple Western 10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:20, Immunohistochemistry-Paraffin 1:10-1:500
Gene Accession No. Q9UHD2
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK NAK serine NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK TANK binding kinase TANK-binding kinase 1NF-kB-activating kinase, TBK 1
Gene Symbols TBK1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 563-577 (YYQFKKDKAERRLAYC) of human TBK1 was used as immunogen for this antibody (NP_037386).
Molecular Weight of Antigen 84 kDa
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29110
Test Specificity Cross reacts with Cow, Dog, Human, Mouse and Rat. Predicted to react with Chimpanzee.
Target Species Bovine, Mouse, Canine, Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.