Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBK1 Antibody (14D10), Novus Biologicals™
SDP

Catalog No. NB298393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298393 100 μL
NB298392 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298393 Supplier Novus Biologicals Supplier No. NBP326292100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TBK1 Monoclonal antibody specifically detects TBK1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry

Specifications

Antigen TBK1
Applications Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 14D10
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human TBK1 [UniProt Q9UHD2]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29110
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.