Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBK1 Antibody (14D10), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | TBK1 |
---|---|
Clone | 14D10 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
TBK1 Monoclonal antibody specifically detects TBK1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
TBK1 | |
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1 | |
A synthesized peptide derived from Human TBK1 [UniProt Q9UHD2] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
14D10 | |
Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Phospho Specific, Protein Kinase, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
29110 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title