Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBLR1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB60270A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60270A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60270A532 Supplier Novus Biologicals Supplier No. NB600270AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBLR1 Polyclonal specifically detects TBLR1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TBLR1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C21, DC42, F-box-like/WD repeat-containing protein TBL1XR1, FLJ12894, IRA1Transducin beta-like 1X-related protein 1, nuclear receptor co-repressor/HDAC3 complex subunit, Nuclear receptor corepressor/HDAC3 complex subunit TBLR1, TBLR1TBL1-related protein 1, transducin (beta)-like 1 X-linked receptor 1, transducin (beta)-like 1X-linked receptor 1
Gene Symbols TBL1XR1
Host Species Rabbit
Immunogen A synthetic peptide that maps to a region between residues 100 and 150 of human TBL1-related Protein 1 using the numbering given in SwissProt entry Q9BZK7 (GeneID 79718).
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 79718.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.