Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBLR1 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB600270IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600270IR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600270IR Supplier Novus Biologicals Supplier No. NB600270IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBLR1 Polyclonal specifically detects TBLR1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TBLR1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C21, DC42, F-box-like/WD repeat-containing protein TBL1XR1, FLJ12894, IRA1Transducin beta-like 1X-related protein 1, nuclear receptor co-repressor/HDAC3 complex subunit, Nuclear receptor corepressor/HDAC3 complex subunit TBLR1, TBLR1TBL1-related protein 1, transducin (beta)-like 1 X-linked receptor 1, transducin (beta)-like 1X-linked receptor 1
Gene Symbols TBL1XR1
Host Species Rabbit
Immunogen A synthetic peptide that maps to a region between residues 100 and 150 of human TBL1-related Protein 1 using the numbering given in SwissProt entry Q9BZK7 (GeneID 79718).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 79718
Test Specificity NB600-270 is specific for human TBLR1 protein.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.