Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tbp7 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB324419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324419 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324419 Supplier Novus Biologicals Supplier No. NBP299612MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tbp7 Polyclonal antibody specifically detects Tbp7 in Human samples. It is validated for Western Blot

Specifications

Antigen Tbp7
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias MB67 interacting protein, MGC13687,26S protease regulatory subunit 6B, MGC23214, MGC8570, protease 26S subunit 6, proteasome (prosome, macropain) 26S subunit, ATPase, 4,26S proteasome AAA-ATPase subunit RPT3, Proteasome 26S subunit ATPase 4, S6Tat-binding protein 7, TBP-7MB67-interacting protein, TBP7MIP224
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Tbp7.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5704
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.