Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBR-2 Rabbit anti-Mouse, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $452.71
Specifications
Antigen | TBR-2 |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN1687-25UG
|
MilliporeSigma
ABN168725UG |
25 μg |
Each for $192.35
|
|
|||||
ABN1687-100
|
MilliporeSigma
ABN1687100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-TBR-2, Cat. No. ABN1687, is a highly specific rabbit polyclonal antibody that targets Eomesodermin homolog (Tbr2) and has been tested in Immunohistochemistry (Paraffin), Immunofluorescence, and Western Blotting.Specifications
TBR-2 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Epigenetics & Nuclear Function | |
Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide. | |
NP_034266 | |
Conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of murine TBR-2. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
Eomesodermin homolog;T-box brain protein 2;T-brain-2 | |
Eomes;Tbr2 | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody detects T-box brain protein 2 (TBR-2) in murine species. It targets an epitope with in 20 amino acids from the C-terminal region. | |
∽63 kDa observed; 74.80 kDa calculated. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title