Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBRG4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP258726 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP258726 100 μL
NB393306 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP258726 Supplier Novus Biologicals Supplier No. NBP258726
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBRG4 Polyclonal specifically detects TBRG4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TBRG4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias cell cycle progression 2 protein, Cell cycle progression protein 2, Cell cycle progression restoration protein 2, CPR2, FAST kinase domains 4, FASTKD4Cpr2, KIAA0948H_TD2522F11.8, protein TBRG4, transforming growth factor beta regulator 4FAST kinase domain-containing protein 4
Gene Symbols TBRG4
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FHQLLCLLNSQIASVWHGTLSKLLGSLYALGIPKASKELQSVEQEVRWRMRKLKYKHLAFLAESCATLSQEQHSQELLAELLTHLERRWTEIEDSHTLVTVMMKVG
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9238
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.