Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
General Data Healthcare™ Flotation Work Station Bath
Shop All Triangle Biomedical Sciences Inc. (TBS) Products

Click to view available options
Amperage:
1.75 A
2.5 A
3.5 A
5 A
Type:
Flotation Work Station
Tissue Flotation Bath
Model:
FWS-L
FWS-S
TFB-L
TFB-S
Description
Ergonomically designed to provide reliability with advanced digital microprocessor control over temperature protection for preparation of paraffin embedded tissue sections
Combined with General Data’s histology consumable and reagents, these baths ensure whole ribbon sections are properly collected without cracking or over-expansion
Basic Bath
- Bath control panel features a keypad and two LED displays
- Arrow keys let you set and display water temperature from ambient up to 65°C (149°F)
- Microprocessor-controlled components heat bath quickly and evenly (ambient to 65°C [149°F])
- Second LED display switches between actual water temperature and time of day
- Magnetic, noncontact switch functions flawlessly for years
- Includes everything in the basic bath, and also integrates a HISTO/Orientator™ heating block and slide dryer
- The 2 × 2 in. (5 × 5cm) heating block can be used after removing slide from the bath to eliminate any remaining bubbles in the tissue
- The 2 × 8 in. (5 × 20cm) slide dryer can dry several slides simultaneously
- Both heating block and slide dryer located above control panel so as not to interfere with bath
Order Info
Accessories are available (See Cat. Nos. 22-047-239, -241, -242 and 22-044-630).
Specifications
Specifications
Controller Type | Microprocessor |
Certifications/Compliance | UL, CSA, CE |
Type | Tissue Flotation Bath |
Display | LED |
Includes | 11 x 8 in. Removable Glass bowl with built-in clock and LED light array |
Model | TFB-L |
Dimensions (L x W x H) | 18.5 x 14 x 5.4 in. (47 x 36 x 14 cm) |
Length (English) | 18.5 in. |
Length (Metric) | 47 cm |
Width (English) | 14 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction