Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | TBX3 |
---|---|
Concentration | 500 μg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119454
|
Invitrogen™
PA5119454 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121561
|
Invitrogen™
PA5121561 |
20 μL |
Each for $209.50
|
|
|||||
Description
TBX3 Polyclonal Antibody for Western Blot
T-box 3 (TBX3) is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. The Tbx3 protein is a transcriptional repressor that binds the canonical Brachychury bindig sites, and is thought to play a role in the anterior/posterior axis of the tetrapod forelimb. Tbx3 is required for normal mammary development and is also implicated in tumor development. Tbx3 expression also increases during osteoblast differentiation, and may function as a determinant of osteoblast cell numbers. Mutations in T-box 3 cause ulnar-mammary syndrome (MIM 181450), affecting limb, apocrine gland, tooth, hair, and genital development. Alternative splicing results in three transcript variants encoding different isoforms; however, the nature of one full length variant has not been determined. Subsets of human breast cancer cell lines overexpress TBX36. in vitro, Tbx3 overexpression in mouse embryo fibroblasts leads to immortalization of cells. Truncated forms of Tbx3 are increased in plasma samples.Specifications
TBX3 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Tbx3 | |
bladder cancer related protein XHL; D5Ertd189e; T-box 3; T-box 3 (ulnar mammary syndrome); T-box protein 3; T-box transcription factor TBX3; TBX3; TBX3-ISO; UMS; XHL | |
Tbx3 | |
Primary | |
Antigen affinity chromatography |
500 μg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
O15119 | |
6926 | |
Recombinant protein T-Box Protein 3. The antigen corresponds to amino acid range 107-220 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title