Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC AQP1 1
GREENER_CHOICE

Catalog No. p-5546196
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Aquaporin 1 (AQP1) channel blocker

TC AQP1 1 is an aquaporin 1 (AQP1) channel blocker (IC50 = 8 μM). Inhibits water flux in Xenopus oocytes.

Specifications

Quantity 10 mg
Product Type TC AQP1 1
Purity >98%
Molecular Weight (g/mol) 218.211
Inhibitors TC AQP1 1

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.