Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC ASK 10
GREENER_CHOICE

Catalog No. 482510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-251-0 10 mg
48-255-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-251-0 Supplier Tocris Bioscience™ Supplier No. 4825/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ASK1 inhibitor; orally bioavailable

Potent ASK1 inhibitor (IC50 = 14 nM); displays selectivity for ASK1 over other kinases including ASK2 (IC50 = 0.51 μM), MEKK1, TAK1, IKKβ, ERK1, JNK1, p38α, GSK-3β, PKCθ and B-raf (IC50 values are > 10 μM). Blocks downstream JNK1/p38 phosphorylation in cells. Orally bioavailable.

Chemical Identifiers

CAS 1005775-56-3
Molecular Formula C21H23Cl2N5O

Specifications

Quantity 10 mg
Formula Weight 432.35
Purity >98%
Molecular Weight (g/mol) 432.349
Chemical Name or Material TC ASK 10
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.