Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-DAPK 6
GREENER_CHOICE

Catalog No. p-4817293
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective inhibitor of DAPK1

Potent and selective, ATP-competitive inhibitor of death-associated protein kinase 1 (DAPK1) (IC50 values are 69 and 225 nM for DAPK1 and DAPK3 respectively, when assayed with 10 μM ATP). Displays selectivity for DAPK1 over a range of 48 other kinases, including Abl, AMPK, Chk1, Met and Src (IC50 > 10 μM).

Chemical Identifiers

CAS 315694-89-4
Molecular Formula C17H12N2O2

Specifications

Quantity 10 mg
Formula Weight 276.29
Purity >98%
Molecular Weight (g/mol) 276.295
Chemical Name or Material TC-DAPK 6
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.