Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TC-FPR 43

Click to view available options
Quantity:
10 mg
50 mg
Description
TC-FPR 43 is a potent formyl peptide receptor 2 (FPR2) agonist (EC50 = 44 nM). Inhibits fMLP- and IL-8-induced neutrophil migration. Suppresses inflammation in a mouse model of acute inflammation. Orally bioavailable.
Specifications
Specifications
Quantity | 10 mg |
Product Type | TC-FPR 43 |
Purity | >98% |
Molecular Weight (g/mol) | 384.86 |
Inhibitors | TC-FPR 43 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction