Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-G 24
GREENER_CHOICE

Catalog No. 435310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-531-0 10 mg
43-535-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-531-0 Supplier Tocris Bioscience™ Supplier No. 4353/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective GSK-3β inhibitor

Potent GSK-3β inhibitor (IC50 = 17 nM); displays selectivity for GSK-3β over CDK2 (22% inhibition at 10 μM). Increases liver glycogen reserves in rodents. Brain penetrant.

Chemical Identifiers

CAS 1257256-44-2
Molecular Formula C15H11ClN4O3

Specifications

Quantity 10 mg
Formula Weight 330.73
Purity >98%
Molecular Weight (g/mol) 330.728
Chemical Name or Material TC-G 24
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.