Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC LPA5 4
GREENER_CHOICE

Catalog No. 470810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-081-0 10 mg
47-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-081-0 Supplier Tocris Bioscience™ Supplier No. 4708/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

LPA5 receptor antagonist

TC LPA5 4 is an LPA5 receptor antagonist (IC50 = 0.8 μM in LPA5-RH7777 cells). TC LPA5 4 inhibits LPA-induced aggregation of isolated human platelets. Exhibits selectivity for LPA5 against 80 other screened targets.

Chemical Identifiers

CAS 1393814-38-4
Molecular Formula C23H23ClN2O3

Specifications

Quantity 10 mg
Formula Weight 410.89
Purity >98%
Molecular Weight (g/mol) 410.898
Chemical Name or Material TC LPA5 4
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.